Learn More
Thermo Scientific™ Finnpipette™ F1 Variable Volume Pipettes
Maximize user comfort and reduce strain with the F1 system, which delivers performance with precision, all in a lightweight design. It's the ideal system for applications using expensive reagents and valuable samples.
$783.83 - $849.89
Specifications
Description Finnpipette F1
Number of Channels 1
Type Variable Volume Pipette
View More Specs
Products 14
Catalog Number Mfr. No. Volume (Metric) Color Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Color Price Quantity  
14-386-300N
View Documents
Thermo Scientific™
4641010N
0.2 to 2 μL Pink
Each for $849.89
Only null left
14-386-301N
View Documents
Thermo Scientific™
4641020N
0.5 to 5 μL Pink
Each for $783.83
Only null left
14-386-302N
View Documents
Thermo Scientific™
4641030N
1 to 10 μL Pink
Each for $790.00
Only null left
14-386-303N
View Documents
Thermo Scientific™
4641040N
1 to 10 μL Yellow
Each for $849.89
Only null left
14-386-304N
View Documents
Thermo Scientific™
4641050N
2 to 20 μL Turquoise
Each for $783.83
Only null left
14-386-305N
View Documents
Thermo Scientific™
4641060N
2 to 20 μL Yellow
Each for $790.00
Only null left
14-386-306N
View Documents
Thermo Scientific™
4641070N
10 to 100 μL Yellow
Each for $790.00
Only null left
14-386-307N
View Documents
Thermo Scientific™
4641080N
20 to 200 μL Yellow
Each for $790.00
Only null left
14-386-308N
View Documents
Thermo Scientific™
4641090N
30 to 300 μL Orange
Each for $783.83
Only null left
14-386-309N
View Documents
Thermo Scientific™
4641100N
100 to 1000 μL Blue
Each for $790.00
Only null left
14-386-310N
Accessories View Documents
Thermo Scientific™
4641110N
0.5 to 5 mL Green
Each for $790.00
Only null left
14-386-311N
Accessories View Documents
Thermo Scientific™
4641120N
1 to 10 mL Red
Each for $790.00
Only null left
14-386-292N
View Documents
Thermo Scientific™
4641130N
5 to 50 μL Turquoise
Each for $783.83
Only null left
14-386-293N
View Documents
Thermo Scientific™
4641140N
5 to 50 μL Yellow
Each for $843.25
Only null left
Description
Description
Enjoy the enhanced volume adjustment on Thermo Scientific™ Finnpipette™ F1 Variable Volume Pipettes, which is now silent and effortless with textured adjustment knob for a firmer feel when setting volumes. The adjustable finger rest and lightweight design maximize comfort without sacrificing performance
For decades, Thermo Scientific Finnpipette pipetting systems have delivered unmatched productivity and ergonomics across thousands of labs and applications worldwide.
Finnpipette F1: Everything you want in your everyday, go-to pipette
Ergonomically In-touch
Lightweight handle design combined with low plunger, tip attachment and ejection forces help reduce the risk of RSI
Silent, Effortless Volume Adjustment
Enjoy the new silent and lighter volume adjustment mechanism with over 50% lower forces compared to earlier models.
Secure Volume Lock
The secure volume setting prevents undesirable volume drift during operation. Simply lift the plunger up to set volume and press down to lock in place with minimum effort.
Adjustable Comfort
The 120° adjustable finger rest – for both right- and left-handed operator comfort – allows the hand to relax during pipetting cycles.
Super Blow-out
150% increase in air boost to ensure efficient delivery of micro-volumes and prevent capillary action in 50μl models and below.
Wide Selection of Finntip Pipette Tips
Finntips are designed and manufactured expressly for Finnpipette models, enabling optimal performance, precision and accuracy.
5 Year Warranty
With convenient online registration, and may vary by country.
Learn more about the complete Finnpipette family >
Specifications
Specifications
Finnpipette F1
1
Variable Volume Pipette
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'