Learn More
Thermo Scientific™ Finnpipette™ F2 Variable Volume Pipettes
Meet the challenges of highly demanding applications where robustness and durability are essential with the sleek, rugged design of these fully autoclavable pipettes.
$840.96 - $915.34
Specifications
| Description | Finnpipette F2 |
|---|---|
| Autoclavable | Autoclavable |
| Number of Channels | 1 |
| Product Line | Finnpipette™ F2 |
| Type | Variable Volume Pipette |
| Catalog Number | Mfr. No. | Volume (Metric) | Compatible Tips | Color | Increments | Price | Quantity | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Volume (Metric) | Compatible Tips | Color | Increments | Price | Quantity | ||
|
14-386-312
|
Thermo Scientific™
4642010 |
0.2 to 2 μL | Finntip 10 Micro, Filter 10 Micro, Flex 10, Flex Filter 10 | Pink | 0.002μL |
Each for $851.82
|
|
||
|
14-386-313
|
Thermo Scientific™
4642020 |
0.5 to 5 μL | Finntip 10 Micro, Filter 10 Micro, Flex 10, Flex Filter 10 | Pink | 0.01μL |
Each for $845.17
|
|
||
|
14-386-314
|
Thermo Scientific™
4642030 |
1 to 10 μL | Finntip 10 Micro, Filter 10 Micro, Flex 10, Flex Filter 10 | Pink | 0.02μL |
Each for $881.19
|
|
||
|
14-386-315
|
Thermo Scientific™
4642040 |
1 to 10 μL | Finntip 250, Filter 10, Flex 200, Flex Filter 30, 200 Ext, Wide 250 | Yellow | 0.02μL |
Each for $840.96
|
|
||
|
14-386-316
|
Thermo Scientific™
4642050 |
2 to 20 μL | Finntip 50 Micro, Filter 50 Micro | Turquoise | 0.02μL |
Each for $845.17
|
|
||
|
14-386-317
|
Thermo Scientific™
4642060 |
2 to 20 μL | Finntip 250, Filter 20, Flex 200, Flex Filter 30, 200 Ext, Wide 250 | Yellow | 0.02μL |
Each for $851.82
|
|
||
|
14-386-318
|
Thermo Scientific™
4642070 |
10 to 100 μL | Finntip 250, Filter 100, Flex 200, Flex Filter 100, 200 Ext, Filter 100 Ext, Wide 250 | Yellow | 0.2μL |
Each for $851.82
|
|
||
|
14-386-319
|
Thermo Scientific™
4642080 |
20 to 200 μL | Finntip 250, Filter 200, Flex 200, Flex Filter 200, 200 Ext, Filter 200 Ext, Wide 250 | Yellow | 0.2μL |
Each for $846.83
|
|
||
14-386-320
|
Thermo Scientific™
4642090 |
100 to 1000 μL | Finntip 1000, Filter 1000, Flex Filter 1000, 1000 Ext, Filter 1000 Ext, Wide 1000 | Blue | 1μL |
Each for $851.82
|
|
||
|
14-386-321
|
Thermo Scientific™
4642100 |
0.5 to 5 mL | Finntip 5mL, Filter 5mL | Green | 0.01mL |
Each for $851.82
|
|
||
|
14-386-322
|
Thermo Scientific™
4642110 |
1 to 10 mL | Finntip 10mL, Filter 10mL, Flex 10mL, Flex Filter 10mL Ext | Red | 0.02mL |
Each for $847.58
|
|
||
|
14-386-294
|
Thermo Scientific™
4642120 |
5 to 50 μL | Finntip 50 Micro, Filter 50 Micro | Turquoise | 0.1μL |
Each for $915.34
|
|
||
|
14-386-295
|
Thermo Scientific™
4642130 |
5 to 50 μL | Finntip 250, Filter 100, Flex 200, Flex Filter 100, 200 Ext, Filter 100 Ext, Wide 250 | Yellow | 0.1μL |
Each for $846.83
|
|
||
Description
Meet the challenges of highly demanding applications where robustness and durability are essential with the sleek, rugged design of Thermo Scientific™ Finnpipette™ F2 Variable Volume Pipettes. Built with tough PVDF components that stand up to harsh chemicals and the damaging effects of UV light, these variable volume pipettes are fully autoclavable without disassembling. They feature a wide selection of expressly designed tips for optimal performance, precision and accuracy.
For decades, Thermo Scientific Finnpipette pipetting systems have delivered unmatched productivity and ergonomics across thousands of labs and applications worldwide.
Finnpipette F2: Proven reliable for long-term intensive use
Rugged and Durable
Finnpipette F2 contains tough PVDF components that stand up to harsh chemicals and the damaging effects of UV light. Its rugged design withstands physical use without damage.
Fully Autoclavable
Decontamination of the Finnpipette F2 is simple as there is no need to disassemble the F2 for autoclaving, minimizing disruption and downtime.
Maintenance is Easy
Simply detach the tip cone for efficient daily maintenance or decontamination when not using an autoclave.
Super Blow-out
150% increase in air boost to ensure efficient delivery of micro-volumes and prevent capillary action in 50μL models and below.
Wide Selection of Finntip Pipette Tips
Finntips are designed and manufactured expressly for Finnpipette models, enabling optimal performance, precision and accuracy.
5 Year Warranty
With convenient online registration, and may vary by country.
Specifications
| Finnpipette F2 | |
| 1 | |
| Variable Volume Pipette |
| Autoclavable | |
| Finnpipette™ F2 |