Thermo Scientific™ Finnpipette™ Novus Multichannel Pipettes
Speed up repetitive dispensing into microplates with these lightweight, multichannel pipettes, with balanced weight distribution for reduced muscle stress and strain.
Supplier:  Thermo Scientific™ 46300400
 View more versions of this product
Catalog No. 14-386-03
Explore available promotions
Description
Description
Speed up repetitive dispensing into 96- and 384-microplates with Thermo Scientific™ Finnpipette™ Novus Multichannel Pipettes. Available in a wide range of models for common laboratory tasks, this product has a lightweight handle and unique index-finger operation that enable longer work periods without fatigue. Simplicity, comfort and functionality — the Novus will be your new best friend in the lab.
Features:
  • Easy-to-read and intuitive menu navigation
  • Multilingual user interface options
  • 10 pipetting options include forward, reverse, diluting, stepper pipetting, mixing, sequential stepping and more
    • 9 aspirate/dispense speeds 
    • Save up to 9 programs with your most common protocols
    • 8 multichannel versions, including a 16-channel for 384-well microplate applications, with color coding by volume for easy recognition
    • Full-size, backlit screen eliminates surrounding light reflections and improves contrast in low-light conditions
    • Quick in-lab calibration for accuracy and precision
    • Fully autoclavable tip cone ensures sterility and avoids cross-contamination
    • Long-life battery allows approximately 4000 pipetting operations; recharged in about one hour
Ergonomic Design
  • Extremely lightweight construction
  • Trigger-action pipetting — index finger controls dispensing with just a soft press of a button
  • 120° adjustable finger rest for balance and comfort
  • Soft-touch tip ejection minimizes thumb strain; can be operated by either hand
Compatible with:
Ideal for use with Thermo Scientific Finntip™ Flex or Finntip pipette tips
Includes:
Finnpipette Novus, universal charger, piston removal tool, instructions for use and calibration certificate
Specifications
Specifications
5.0% to 1.0±
Autoclavable
Flex 300, 300
8
Yes
Multi-Channel Pipette
1µL
Flex 300, 300
Orange
FinnPipette™ Novus
FinnPipette™ Novus
Sterile
30 to 300 μL
2.0 to 0.3%
Videos
SDS
Documents
Documents
Accessories