Thermo Scientific™ Finnpipette™ Stands
Provide safe and convenient storage for manual and electronic pipettes using these stands, with options for single stand, F-stand, multi-stand, multichannel and carousel.
$116.36 - $994.06
Catalog Number | Mfr. No. | Holds | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Holds | Price | Quantity | |||||
14-245-160
|
Thermo Scientific™
9420400 |
Manual Pipettes (6) |
Each for $308.95
|
|
|||||
14-387-14
|
Thermo Scientific™
9420290 |
Manual Pipettes (6) |
Each for $282.09
|
|
|||||
14-387-15
|
Thermo Scientific™
9420320 |
Manual Pipettes (3) |
Each for $222.98
|
|
|||||
21-377-002
|
Thermo Scientific™
9420340 |
Pipettes (6 Manual or 3 Electronic) |
Each for $994.06
|
|
|||||
14-387-21
|
Thermo Scientific™
9420360 |
Electronic Pipette (1) |
Each for $357.33
|
|
|||||
14-245-161
|
Thermo Scientific™
9420390 |
Multichannel Manual Pipette (1) |
Each for $116.36
|
|
|||||
Description
Provide safe and convenient storage for manual and electronic pipettes using Thermo Scientific™ Finnpipette™ Stands. Available options include options for single stand, F-stand, multi-stand, multichannel and carousel.
Finnpipette F-Stand
- Accommodates six pipettes
- Sturdy crossbar structure with slot for each pipette for secure storage
- Suitable for both single channel and multichannel models
Finnpipette Multichannel Stand
- Multi-position stand
- Can hold any multichannel Finnpipette
- Pipette can be stored in upright or horizontal position
- Small footprint
Finnpipette Carousel Stand
- Rotating, portable
- Transparent lid protects pipettes from contamination
- Shipped with three adapters for electronic Finnpipette Novus pipettes and five adapters for manual Finnpipette pipettes
Finnpipette Novus Stand
- For convenient storage of electronic Finnpipette Novus pipette
- Accommodates both single and multichannel Finnpipette Novus models with ease