missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Finntip™ Multistepper Pipette Tips

Perform positive displacement applications with these tips, which are effective for fragile cell suspensions and applications that require outstanding biological purity.

 View more versions of this product

Catalog No. 21-377-34


Explore available promotions

Description

Description

Perform applications that demand the utmost in biological purity; positive displacement applications; and pipetting fragile cell suspensions and macromolecules with Thermo Scientific™ Finntip™ Multistepper Pipette Tips and the Finnpipette Multistepper pipette. Available both as sterile and non-sterile.

Features:

  • For use with Thermo Scientific Finnpipette™ Multistepper Pipetter
  • Length: 8.8cm
  • Volume: 1500μL
Specifications

Specifications

Finntip Multistepper, 10x96/rack, nonsterile
Non-filtered
Turquoise
8.8cm
Rack
FinnTip
1500 μL
Non-sterile
3.46 in.
ISO 9001, ISO 14001
Thermo Scientific FinnTip Multistepper
Videos
SDS
Documents

Documents

Promotions

Promotions