missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisher BioReagents™ Microbiology Media Additives: Yeast Extract
Additive for preparing bacterial nutrient media
$95.16 - $599.21
Specifications
Form | Dehydrated |
---|---|
Grade | Molecular genetics |
Media Type | Yeast Extract |
Product Type | Microbiology Media |
Packaging Type | Poly Bottle |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
BP1422-100
|
Fisher BioReagents™
BP1422100 |
100 g |
N/A
|
|
|||||
BP1422-500
|
Fisher BioReagents™
BP1422500 |
500 g |
N/A
|
|
|||||
BP1422-2
|
Fisher BioReagents™
BP14222 |
2000 g |
N/A
|
|
|||||
Description
Water soluble portion of autolyzed fresh yeast used in bacteriological culture mediaSpecifications
Dehydrated | |
Yeast Extract | |
Poly Bottle |
Molecular genetics | |
Microbiology Media |