Fisherbrand™ Bacti-Loop™ Micro-Sterilizer
Complete sterilization in seconds without spatter
Supplier:  Fisherbrand™ B1000FI
Catalog No. 22-630-001
Explore available promotions
Description
Description
Complete sterilization in seconds without spatter
Warranty
One year
Specifications
Specifications
2 A
15 cm
0.55 in.
1.39 cm
16.5 cm
Sterilizer
500°C or 815°C
1.4 kg
50/60 Hz
5 to 7 sec.
4 in.
6 in.
15 cm
6 in.
6.5 in.
22630001
1500°F
2.9 lb.
120 V 50/60 Hz
Stainless Steel
120 V
10 cm