missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisher Healthcare™ PROTOCOL™ Hema-G Stains for Geometric Data Hemastainer™
Quality-control tested on human blood smears to ensure proper staining and absence of precipitate
$359.71 - $672.55
Specifications
Storage Requirements | Room Temperature |
---|---|
For Use With (Equipment) | Geometric data Hemastainer™ |
Catalog Number | Mfr. No. | Quantity | Type | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Type | Price | Quantity | |||||
23-011114
|
Fisher Healthcare™
23011114 |
1 gal. | Hema-G Wright Stain, New Formulation |
Each for $386.77
Case of 4 Each for $1,547.08
|
|
|||||
23-011115
![]() |
Fisher Healthcare™
23011115 |
1 gal. | Hema-G Wright-Giemsa Stain, New Formulation |
Each for $400.35
Case of 4 Each for $1,601.40
|
![]() |
|||||
23-247057
|
Fisher Healthcare™
247057 |
1 gal. | Hema-G Wright-Giemsa Stain, Regular |
Each for $672.55
Case of 4 Each for $2,690.20
|
|
|||||
23-247056
|
Fisher Healthcare™
247056 |
1 gal. | Hema-G Wright Stain, Regular |
Each for $527.58
Case of 4 Each for $2,110.32
|
|
|||||
23-247058
|
Fisher Healthcare™
247058 |
1 gal. | Hema-G Liquid Buffer |
Each for $359.71
Case of 4 Each for $1,438.84
|
|
|||||
Description
Stains for manual and automated staining methods
- All PROTOCOL Hematology Stains are prepared with materials certified by the Biological Stain Commission to ensure purity
- Manufactured with strict quality control measures to ensure lot-to-lot uniformity, including extensive analytical and performance testing on human specimens
- Carefully balanced for optimum staining pH
- Remain stable for a minimum of 12 months when stored at room temperature
- Allow varied staining times to suit individual preferences
- Packaged in durable plastic bottles for safety and easy handling
- Convenient finger tabs and clear reagent identification eliminate improper line connection
Specifications
Room Temperature | |
Geometric data Hemastainer™ |