missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisher Healthcare™ PROTOCOL™ Histological/Cytological Stains

Manufactured with strict quality control to ensure lot-to-lot uniformity

$92.35 - $458.00

Specifications

Format Liquid
For Use With (Application) Used for Histological/Cytological Staining
View More Specs

Products 16
Catalog Number Mfr. No. Type Quantity Price Quantity  
Catalog Number Mfr. No. Type Quantity Price Quantity  
23-245651
View Documents
Fisher Healthcare™
245651
Harris Hematoxylin - Mercury-free 4L
Each for $355.29
Case of 4 Each for $1,421.16
 
23-245655
View Documents
Fisher Healthcare™
245655
Gill Hematoxylin (2X) 4L
Each for $426.81
Case of 4 Each for $1,707.24
 
23-245656
View Documents
Fisher Healthcare™
245656
Gill Hematoxylin (3X) 475mL
Each for $96.29
Case of 12 Each for $1,155.48
 
23-245653
View Documents
Fisher Healthcare™
245653
Gill Hematoxylin (1X) 4L
Each for $402.21
Case of 4 Each for $1,608.84
 
23-245658
View Documents
Fisher Healthcare™
245658
Alcoholic Eosin Y Stain 1L
Each for $136.42
Case of 12 Each for $1,637.04
 
23-245657
View Documents
Fisher Healthcare™
245657
Gill Hematoxylin (3X) 4L
Each for $458.00
Case of 4 Each for $1,832.00
 
23-245654
View Documents
Fisher Healthcare™
245654
Gill Hematoxylin (2X) 475mL
Each for $208.68
Case of 12 Each for $2,504.16
 
23-245681
View Documents
Fisher Healthcare™
245681
Bluing Reagent 4L
Each for $215.81
Case of 4 Each for $863.24
 
23-245678
View Documents
Fisher Healthcare™
245678
Harris Hematoxylin 4L
Each for $454.06
Case of 4 Each for $1,816.24
 
23-245677
View Documents
Fisher Healthcare™
245677
Harris Hematoxylin- Mercury-free 475mL
Each for $92.35
Case of 12 Each for $1,108.20
 
23-245679
View Documents
Fisher Healthcare™
245679
Gomori Trichrome 500mL
Each for $208.87
Case of 12 Each for $2,506.44
 
23-245697
View Documents
Fisher Healthcare™
245697
Harris Hematoxylin - Mercury-free, Non-acidified 4L
Each for $414.74
Case of 4 Each for $1,658.96
 
23-245827
View Documents
Fisher Healthcare™
245827
Alcoholic Eosin Y Stain 4L
Each for $272.26
Case of 4 Each for $1,089.04
 
23-310349
View Documents
Fisher Healthcare™
310349
Cyto-Quik Combo EA/OG Stain 1 gal.
Each for $433.90
Case of 4 Each for $1,735.60
 
23-314631
View Documents
Fisher Healthcare™
314631
Eosin Y Intensified 1 gal.
Each for $264.23
Case of 4 Each for $1,056.92
 
23-314630
View Documents
Fisher Healthcare™
314630
Eosin Y Intensified 32 oz.
Each for $136.16
Case of 12 Each for $1,633.92
 
Description

Description

Prepared with stain certified by the Biological Stain Commission

Specifications

Specifications

Liquid
Used for Histological/Cytological Staining
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'