Fisher Healthcare™ Sure-Vue™ Adeno Test Kit
Simple two-step procedure
$404.53 - $404.53
Specifications
CPT Code | 87301 |
---|---|
For Use With (Application) | Qualitative detection of Adenovirus and its Antigens in eye swabs, nasal and pharyngeal secretions and fecal material |
Includes: 20 Test devices, tubes containing extraction buffer, disposable sample transfer pipets, package insert
Catalog Number | Mfr. No. | Quantity | Type | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Type | Price | Quantity | ||||
SA067020
|
Fisher Healthcare™
067020F |
20 Tests | Adenovirus Test Kit |
Pack of 20 Each for $404.53
|
|
||||
Description
- CLIA Complexity: Moderate
- CPT Code: 87301
- Results in 15 minutes
- Rapid, membrane-based immunogold assay for the qualitative detection of adenovirus and its antigens
- Aids in the diagnosis of conjunctivitis, respiratory illnesses and gastroenteritis caused by adenovirus
- Can be used with eye swabs, nasopharyngeal secretions, fecal samples, and cell culture supernatant
- Positive and negative controls sold separately
Specifications
87301 | |
Qualitative detection of Adenovirus and its Antigens in eye swabs, nasal and pharyngeal secretions and fecal material |