missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisher Healthcare™ Sure-Vue™ RSV Test Kit
Alternative procedure for bloody and mucoid samples
Supplier: Fisher Healthcare™ 041130F
Includes: 30 Test devices, extraction tubes with filter caps, two bottles extraction buffer (10mL each), disposable pipettes, quick reference card and package insert
Description
- CLIA Complexity: Waived and Moderate
- CPT Codes: 87807QW, 87807
- Sensitivity: >99%
- Specificity: >99%
- Results in 15 minutes
- Specimen types: Nasopharyngeal washes, aspirates and swabs
- Built-in procedural control
Specifications
87807, 87807QW | |
Respiratory Syncytial Virus (RSV) | |
RSV Test Kit | |
30 Tests |
Waived, Moderate | |
Nasal Aspirate, Nasal Swab, Nasal Wash | |
30 Test devices, extraction tubes with filter caps, two bottles extraction Buffer, disposable pipettes, quick reference card and package insert | |
15 min. |