Fisherbrand™ Faucet-Mount Personal Eyewash
Outfit your workspace with the Fisherbrand Faucet-Mount Personal Eyewash, and ideal eyeash station for laboratory applications.
Supplier: Fisherbrand™ S19200B0470
Includes: Safety sign and inspection tag
Description
Unit is activated by an actuator pin on adapter. When pin is in closed position, faucet can be used for normal operation.
- Mounts to both standard and gooseneck faucets
- Features chrome-plated brass pipe, fittings, faucet adapter, and twin sprayhead assembly
- Yellow dust covers pop off when eyewash is activated
To eliminate risk of scalding the eyes, disconnect hot water supply to the faucet or install tempered water system.
Order Info
Adapter on unit is 0.86 in. x 27 thread (female), but includes two additional adapters to fit most faucets (0.81 in. x 27 thread [male] and 0.94 in. x 27 thread [male])
Certifications
SEI certified to comply with ANSI™ Z358.1-2004.
Specifications
Actuator Pin on Adapter | |
Standard and Gooseneck Faucets for Laboratory Applications | |
Faucet |
ANSI Z358.1-2004, SEI | |
Twin sprayhead, Chrome Plated Brass pipe, fittings, faucet adapter, yellow pop-off dust cap, universal identification sign, inspection tag | |
Faucet Mount Personal Eyewash |