missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Hazard and NFR Identification Write-On Labels
Ideal for adding vital hazard information to surfaces of small bottles, cans, and other containers.
Supplier: Fisherbrand™ J066536
Description
Blank hazard classification diamond can be filled in with the appropriate ratings.
Large Labels
- Spaces for Chemical Name, Hazards and Precautions
- Check boxes for goggles, gloves, apron and fume hood
- Dimensions: 5W × 3 in.H (12.7 × 7.6cm)
- Spaces for Chemical Name, Owner and Date
- Dimensions: 3.88W × 1.5 in.H (9.9 × 3.8cm)
Specifications
Label | |
Chemical hazard | |
5 in. | |
7.6 cm |
Hazard Identification Label | |
100/Roll | |
3 in. |