missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 24-Hour Timer/Clock

With big digit display.

Supplier:  Fisherbrand™ 7898

Catalog No. S01621

Only null left
Explore available promotions

Description

Description

  • Max./Min. count up/down
  • Memory for preset value
  • 12/24 hours format
  • Four timer count up/down
  • Foldaway stand
  • Magnetic attachment

Specifications:

  • Maximum count down/up range: 99 hours 59 minutes 59 seconds
  • Battery: 1.5V, size AAA x 1 pc
  • Display size: 66 x 22mm
  • Digit size: 10 x 18mm
Specifications

Specifications

Clock/Timer
Digital
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions