accumet™ Electrode Storage Solution
Supplier:  Thermo Fisher Scientific BPSE401
Catalog No. SE40-1
Explore available promotions
Description
Description
Solution of potassium hydrogen phthalate—potassium chloride conditions electrodes when not in use
Specifications
Specifications
100°C
1000 mL
Electrode
Miscible
33.8 oz.
Clear
Storage Solution