Fisherbrand™ accumet™ pH Indicating Glass Body Universal Glass Membrane Models- Mercury-Free
Universal glass membrane and glass body provides measurements from zero to 14pH
$566.48 - $566.48
Catalog Number | Mfr. No. | Connector Types | Electrode | Length (English) | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Connector Types | Electrode | Length (English) | Price | Quantity | |||
13-620-3
|
Fisherbrand™
136203 |
U.S. Standard | Suggested Reference Electrodes: 13-620-51, 13-620-52, 13-620-57, 13-620-61, 13-620-62, 13-620-45, 13-620-53, 13-620-46, 13-620-47 | 30 in. |
Each for $566.48
|
|
|||
Description
Universal glass membrane and glass body provides measurements from zero to 14pH
- Universal glass membrane
- Zero to 14pH range
Specifications
Glass | |
0 to 14 |