Fisherbrand™ accumet™ Probe
Simple Plug and Stir technology for accumet XL and AB series meters
Supplier:  Fisherbrand™ 13620BSP
Catalog No. 13-620-BSP
Explore available promotions
Description
Description
Simple Plug and Stir technology for accumet XL and AB series meters
Provides mixing without magnetic stir bars or stir plates required.
  • Saves valuable bench space
  • Quiet operation
  • Adjustable speed control
  • No additional power required
  • Multi-parameter XL benchtop models can accept up to two probes for simultaneous stirring
Order Info
Compatible with the following models: XL150, XL200, XL250, XL500, XL600, AB150, AB250, AB200. Replacement paddles available separately; see cat. no. 13620RP.
Warranty
One year
Specifications
Specifications
Probe
Model XL150, 200, 250, 500, 600, AB150, 200 and 250 Multiparameter Benchtop Meter