missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ accuWash™ and accuWash Versa Microplate Washers
Ease of use, convenience and versatility for washing microplates in ELISA applications
$13314.17 - $18712.23
Specifications
Residual Volume | <2 μL |
---|---|
Weight (Metric) | 9 kg |
Dimensions (L x W x H) | 15.2 x 13.6 x 9.4 in. (38.5 x 34.5 x 24 cm) |
Weight (English) | 20 lb. |
Electrical Requirements | 100/240 V 50/60 Hz |
Includes:
accuWash: Includes 1x2L wash bottle, 1x2L waste bottle, 1x8 wash head, aerosol cover
accuWash Versa: Includes 2x2L wash bottle, 2 x 1L rinse bottle, 1x4L waste bottle, 2x8 wash head, aerosol cover
Catalog Number | Mfr. No. | Description | Plate Type | Cell Washing | Wash/Waste Bottle Capacity | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Plate Type | Cell Washing | Wash/Waste Bottle Capacity | Price | Quantity | ||
14-377-577
|
Fisherbrand™
5165090 |
accuWash | 96-Well Plates | No | 1 |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
14-377-578
|
Fisherbrand™
5165100 |
accuWash Versa | 96-well Plate, 384-well Plate | Yes | Up to 3 |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
The Fisher Scientific accuWash and Fisher Scientific accuWash Versa are easy-to-use strip washers that offer convenience through a large color screen, intuitive graphical user interface, and a USB port. The accuWash is the basic model for washing 96-well plates only. It is intended for use when only a few similar assays are run routinely. The accuWash Versa can also wash cells and 384-well plates, offering enhanced flexibility needed for research use.
Key Features
- Easy protocol setup with the large color screen and the visual internal software
- A USB port for transferring protocols between several washers, and for easy updating of the internal software
- Sweep mode provides an extremely low residual volume in the well, resulting in excellent washing performance and reliable assay results
Easy and convenient to use
- Large color screen shows everything needed for normal use on one display
- Internal software is logical and intuitive, and allows protocols to be developed fast, making it easy to get started with the software; requires minimal training
Reliable and secure performance
- Unpressurized bottles are safe and secure to use
- Automatic rinse and prime features prevent clogging of liquid transfer system
- Liquid level sensors provide security for both wash and waste bottles
- Low residual volumes of washers provide optimal washing performance, helping deliver reliable assay results
Added versatility
- Washes cells, 96- and 384-well plates
Certifications
CE mark
Warranty
1 year
Specifications
<2 μL | |
15.2 x 13.6 x 9.4 in. (38.5 x 34.5 x 24 cm) | |
100/240 V 50/60 Hz |
9 kg | |
20 lb. | |
100/240 V |