Fisherbrand™ Air-Powered Analog Overhead Stirrers
Variable speed and explosion-proof stirrers ideal for safely mixing all types of solvents, lacquers and volatile chemicals without the possiblity of spark ignition.
Supplier:  Fisherbrand™ 370209017
 View more versions of this product
Catalog No. 14-500-219
Explore available promotions
Description
Description
  • Needle valve allows variation in mixing speed
  • Special muffler ensures quieter operation
  • Reversible models have motors which can be set to rotate in either direction by connecting the air supply line to the right port for counter-clockwise rotation or the left port for clockwise rotation
  • Can be used with flammable chemicals for explosion proof mixing
Specifications
Specifications
10,000 rpm
0.37 in.
0.3
10 L x 0.56 in. dia. support rod, 4 ft. air hose with double fittings for connection to air supply
45
5 lb.
10 L
0.95 cm
Safely mixing all types of solvents, lacquers and volatile chemicals
30 to 100 psi
5000 mPas
2.3 kg