Fisherbrand™ Digital Thermometers with Stainless-Steel Stem and 0.375 in. LCD Screen
Easy-to-read handheld digital thermometers
Description
Easy-to-read Fisher Scientific™ Digital Thermometers with Stainless-Steel Stem are chemically resistant to acids, bases, solvents, and may be used to measure temperature of gases, food, and other semisolids.
- Short-stem piercing models are ideal for carrying in a pocket
- Longer probes are excellent for measuring sample temperatures while stirring in deep vessels or water baths
- Thermometers with longer sampling rates allow thermometer to be removed from sample before reading—ideal when display is difficult to read in measuring position
- For use in cuvets, soil, incubators, petroleum products and foods
Certifications
Individually serial-numbered Traceable Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™; indicates traceability to standards provided by NIST™.
Specifications
ISO 17025, A2LA, NIST | |
Measuring temperatures of semi solids gunky and viscous solutions | |
5.25 in. | |
13.3 cm | |
1 oz. | |
6.5 in. | |
150°C | |
-50°C | |
-50°C to +150°C | |
Digital LCD |
0.15 in. | |
Silver Oxide battery, Stainless Steel probe | |
165 mm | |
0.1°C | |
28 g | |
302°F | |
-58°F | |
-58°F to +302°F | |
±1°C |