Fisherbrand™ Dry Ice/Liquid Nitrogen Trap
Large cold chamber provides rapid condensation of solvent vapors
Supplier: Fisherbrand™ 0125714
Description
- Protects Vacuum pump from harmful vaporss
- Prevents oil back-streaming
- Dry ice/isopropyl alcohol slurry to produce at a temperature of -79°C or liquid nitrogen (-198°C) may be used as refrigerant
- Accepts 1/2 in. O.D. (12.7mm) tubing
Specifications
Dry Ice/Liquid Nitrogen Trap |