Fisherbrand™ Heat/Cool Thermal Mixer Accessory, Thermal Blocks
This Thermal Block is an accessory for the Heat/Cool Thermal Mixer.
$476.95 - $551.93
Catalog Number | Mfr. No. | Holds | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Holds | Price | Quantity | ||||||
05-450-208
|
Fisherbrand™
270691 |
0.5 mL Microtubes |
Each for $476.95
|
|
||||||
05-450-210
|
Fisherbrand™
270693 |
STD/Deep-well Micro-titer Plates |
Each for $551.93
|
|
||||||
Description
- Interchangeable platforms
- Heating block
- Cooling block
- Holds up-to 96 PCR tubes
Specifications
Thermal Block |