missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ High-Temperature Pressure Data Logger

Designed for use in autoclave validation and mapping

 View more versions of this product

Catalog No. 15-060-100


Explore available promotions

Alert:

Battery Warning: Fire, explosion, and severe burn hazard. Do not short circuit, charge, force over discharge, crush, penetrate, or incinerate. Battery may leak or explode if heated above 150°C (320°F).

Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.


Requires : 15-060-081 USB Docking Station with cradle and software package.

Description

Description

Designed for use in autoclave validation and mapping

Easy to use — simply place it in the docking station (15-060-081), connect to a computer, and the device can be started, stopped or downloaded using the software. Graphical, tabular and summary data is provided for analysis and can be viewed in PSIA, mmHg, bar, Torr and kPa. The data can also be automatically exported to Excel for further calculations.

  • Simple setup and installation; minimal long-term maintenance
  • Rugged device can withstand temperatures up to 140°C
  • Completely submersible (IP68)
  • Built with a precision stainless-steel pressure gauge
  • Can be programmed to take readings as often as once per second (1Hz)
  • Nonvolatile memory can store up to 32,700 measurements
  • N.I.S.T. traceable
  • User replaceable lithium battery
  • Engraved label
  • Software battery life indicator
  • May be used with PC to monitor and record data in real-time
  • Digital calibration through software
  • Programmable immediate start or delay start up to six months
  • Accuracy of ±0.03 bar (±0.435 PSI); can be achieved over wide temperature range, from 20°C to +140°C (68°F to 284°F)
Software Features: Multiple graph overlay, Statistics, Digital calibration, Zoom in/ zoom out, Lethality equations (F0, PU), Mean Kinetic Temperature, Full time zone support, Data annotation, Min./Max./Average lines, Data table view, Automatic report generation, Summary view, Multilingual.
Specifications
  • Pressure Sensor: Semiconductor strain gauge
  • Pressure Range: 0 to 5 bar (0 to 72.5 PSIA)
  • Pressure Resolution: 0.1mBar (0.0015 PSIA)
  • Calibrated Accuracy: ±0.03 bar (±0.435 PSI)
  • (20°C to +140°C/68°F to 284°F)
  • Pressure Response Time: 0.1ms (10 to 90% FSR)
  • Repeatability: ±0.5% FSR; ±0.2% typical
  • Reading Rate: 1 reading every second to 1 every 12 minutes
  • Memory: 32,700 readings; software configurable memory wrap
  • Calibration Interval: One year from factory calibration
  • Battery Type: 3.6V lithium (included)
  • Battery Life: 2 years typical
  • Data Format: PSIA, bar, mbar, atm, Torr, Pa, kPa, MPa, mmHg, inHg, inH2O, mH2O, ft, m
  • Time Accuracy: ±1 min./month at 20°C
  • Computer Interface: USB (15-060-081 interface cable reqd.); 125,000 baud
  • Software: XP SP3/Vista/Windows 7
  • Operating Environment: -20°C to +140°C (-4°F to +284°F), 0 to 100% RH
  • Dimensions: 2 x 1 in. dia. (5.08 x 2.54cm)
  • Enclosure: stainless steel
  • Weight: 3 oz. (85g)

  • Autoclave validation and mapping
  • Canning and cooking pouches
  • Pressurized process reactors and streams
  • Beverage production, including wine and soda

Compliance

CE approved

Specifications

Specifications

High Temperature Pressure Datalogger
Digital Calibration
3.6 V Lithium
CE
Stainless Steel
Semiconductor Strain Gauge
20°C to 140°C
2.54 cm
6 cm
85 g
32,700 Readings
XP SP3, Vista, Windows 7
2 Years
USB
PR140
For autoclave validation and mapping, canning and cooking pouches, pressurized process reactors and streams, beverage production, including wine and Soda
1 in.
2.29 in.
3 oz.
Videos
SDS
Documents

Documents

Promotions

Promotions