Learn More
Fisherbrand™ Isotemp™ Digital Dry Baths/Block Heaters
Interchangeable modular blocks, configurable for a range of applications, increase lab versatility.
Supplier: Fisherbrand™ 88860022
Requires : Unit must be ordered with block(s)
Description
Fisherbrand™ Isotemp Digital Dry Baths/Block Heaters offer a range of configurations with interchangeable modular blocks for a variety of application needs.
Fisherbrand™ Isotemp Digital Dry Baths/Block Heaters offer a range of configurations with interchangeable modular blocks for a variety of application needs. Designed for basic to advanced applications with a maximum temperature of 130°C, a timer and available in 1, 2 and 4 block sizes.
- Digital controls and display of time and temperature
- Advanced internal temperature sensing probe for outstanding temperature accuracy and control
- Precise temperature control with PID circuit
- Temperature calibration enables to offset the temperature to desired value
- Timer allows the user to accurately monitor the heating time
- Wide range of interchangeable aluminum alloy heat blocks provide versatility and allow for easy cleaning and disinfecting
- Powder-coated steel body construction ensures durability
- Built-in over-temperature protection device warrants sample and user safety
- Temperature deviation adjustment
Applications:
Sample preparation, enzyme preservation, enzyme-substrate reactions, DNA amplification, electrophoresis gel degeneration and serum coagulation
Warranty
2-year Thermo Fisher Scientific standard, limited warranty. Contact your sales representative for details.
Specifications
CE, cULus, RoHS | |
Powder-coated Steel | |
US | |
Ambient +5° to 130°C | |
250V 5A | |
Dry bath, US style plug, block lifter | |
≤ ± 1°C | |
2 Year, Replacement | |
12.5 in. | |
7.9 in. | |
3.9 in. | |
7.28 lb. | |
100/120 V, 50/60 Hz | |
50/60 Hz |
PID | |
2 | |
<80% | |
Dry Bath/Block | |
≤ 20 min. 30° to 130°C | |
≤ ± 0.5°C | |
0 to 99:59 min or continuous | |
12.5 x 7.9 x 3.9 in. (318 x 200 x 100 mm) | |
318 mm | |
200 mm | |
100 mm | |
3.3 kg | |
100/120 V |