Fisherbrand™ Maxima™ C Plus Pump Oil
Triple-distilled hydrocarbon pump oil designed to resist breakdown in Fisher Scientific Maxima C Plus pumps.
Supplier: Fisherbrand™ 012574A
Description
Designed to resist breakdown in Fisher Scientific Maxima C Plus pumps
Triple-distilled hydrocarbon oil using severely hydrotreated base stock. Hydrotreating minimizes aromatics and sulfur to give good resistance to sludge and varnish formation over time in corrosive environments. The oil enables pumps to maintain the highest vacuum performance over time. Clear, colorless, and odorless.
Specifications
Pump Oil | |
Maxima™ C plus vacuum pump series | |
1 Ea. | |
0.95 L |
Oil enables pumps to maintain the highest vacuum performance over time | |
Clear | |
1 Qt. (0.25 gal.) |