Fisherbrand™ Maxima™ C Plus Pump Oil
Triple-distilled hydrocarbon pump oil designed to resist breakdown in Fisher Scientific Maxima C Plus pumps.
Supplier:  Fisherbrand™ 012574D
 View more versions of this product
Catalog No. 01-257-4D
Explore available promotions
Description
Description
Designed to resist breakdown in Fisher Scientific Maxima C Plus pumps
Triple-distilled hydrocarbon oil using severely hydrotreated base stock. Hydrotreating minimizes aromatics and sulfur to give good resistance to sludge and varnish formation over time in corrosive environments. The oil enables pumps to maintain the highest vacuum performance over time. Clear, colorless, and odorless.
Specifications
Specifications
Pump Oil
Maxima™ C plus vacuum pump series
1 Ea.
19 L
Oil enables pumps to maintain the highest vacuum performance over time
Clear
5.01 gal.