Fisherbrand™ Maxima C Plus Centering Rings
Centering Ring for use with Maxima C vacuum pumps to join two flanges
$45.55 - $71.43
Catalog Number | Mfr. No. | For Use With | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With | Price | Quantity | |||||
01-257-18A
|
Fisherbrand™
0125718A |
Model M2C and M4C vacuum pump |
Each for $45.55
|
|
|||||
01-257-18B
|
Fisherbrand™
0125718B |
Model M8C, M12C and M16C vacuum pump |
Each for $71.43
|
|
|||||
Description
For use with Model M2C and M4C vacuum pumpSpecifications
Stainless Steel | |
Centering Ring |