Fisherbrand™ Maxima C Plus Hose Adapters
Accessory for M2C and M4C vacuum pump.
$82.55 - $99.06
Specifications
Type Hose Adapter
View More Specs
Products 2
Catalog Number Mfr. No. For Use With (Application) For Use With Price Quantity  
Catalog Number Mfr. No. For Use With (Application) For Use With Price Quantity  
01-257-11B
View Documents
Fisherbrand™
0125711B
20.6 to 22.2mm I.D. tubing M8C, M12C, and M16C
Each for $99.06
01-257-11A
View Documents
Fisherbrand™
0125711A
11.1 to 12.7mm I.D. tubing M2C and M4C
Each for $82.55
Description
Description
  • Waterproof, tight fitting connectors
Specifications
Specifications
Hose Adapter
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'