Fisherbrand™ Microtube Holder for Heavy-duty Vortex Mixer
For use with heavy-duty vortex mixer.
Supplier:  Fisherbrand™ 80863047
 View more versions of this product
Catalog No. 02-216-124
Explore available promotions
Description
Description
  • Holds a range of Micro-Tube sizes and numbers
  • Requires Universal Holder
Specifications
Specifications
1.5 to 2 mL
Heavy-duty vortex mixer
38
Microtube Holder
Foam insert
Universal holder
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Accessories