Fisherbrand™ Model 705 Sonic Dismembrator
Programmable ultrasonic processor with an easy to use touchscreen control and overload protection. Ultra-high power with advanced programming for all liquid processing applications.
Supplier:  Fisherbrand™ FB705110
 View more versions of this product
Catalog No. FB705110
Explore available promotions
Includes: Generator, converter, power cable, wrench set, standard 1/2 in. replaceable tip probe (FB4220) for processing 20-250 mL volumes.
Description
Description
The Fisher Scientific™ Model 705 Sonic Dismembrator is a full-featured, programmable ultrasonic processor with an easy to use touchscreen control and overload protection. Ultrahigh power with advanced programming for DNA shearing/ChIP and all liquid processing applications.
  • Processes 0.2 to 1000 mL
  • Voltage: 110 V,  700 watts
  • The additional power and variety of accessories accommodate the widest range of sample volumes and applications.
  • The 4 tip horn and Cup Horn accessories offer time saving, high throughput solutions to repetitive, individual sample processing
  • Programmable from 1 sec. to 99 hrs.
  • Save up to 10 different programs
  • Pulse mode (1 second to 24 hours)
  • Optional Temperature monitoring
  • Applications Include:Cell disruption, homogenization, ChIP/DNA shearing, nanoparticle dispersion, protein extraction, sonochemistry, soil extraction, mixing
Order Info
Stand not included.
Warranty
Two years
Specifications
Specifications
110 V
20 kHz (Ultrasonic Frequency)
0.2 mL to 1 L
9 in.
18 in.
Generator, converter, 1/2 in. dia. probe, power cable, wrench set. Note: Stand and Clamp shown in picture not included
1 sec. to 99 hrs.
110/117 V, 50/60 Hz
700 W
15 x 18 x 9 in.
15 in.
Ideal for cell disruption, homogenization, chip/DNA shearing, nano particle dispersion, protein extraction, sonochemistry, soil extraction, mixing
FB705
SDS
Documents
Documents
Accessories