Fisherbrand™ Power Hold Universal Clamps for Overhead Stirrers
Can be used for most applications.
Supplier:  Fisherbrand™ 370209046
 View more versions of this product
Catalog No. 14-512-906
Explore available promotions
Description
Description
  • Designed with large knobs for powerful gripping
  • Adjustable stop for speed and safety in set up and operation
  • Made with precision machined aluminum rod for strength and durability
Specifications
Specifications
Stirring stands from 9.5mm to 16mm dia.
Universal