Fisherbrand™ Probes for Model 50 and 120 Sonic Dismembrator
Image shows (left to right): 4423, 4422, 4435.
Supplier:  Fisherbrand™ 4435
 View more versions of this product
Catalog No. FB4435
Explore available promotions
Description
Description
These Fisher Scientific™ Probes are designed for use with Model 50 and Model 120 Sonic Dismembrators.
  • 1/4 in. Probe for Model 50/120 Sonic Dismembrator
  • For processing 10 to 50mL volumes
Specifications
Specifications
Probe
10 to 50 mL
1/4 in.
6 mm
1 probe
120 μm maximum
0.25 in.
0.63 cm
Model 120 sonic dismembrator