Fisherbrand™ Pump Repair Kits
For use with vacuum pumps
Supplier:  Fisherbrand™ 0125717B
 View more versions of this product
Catalog No. 01-257-17B
Explore available promotions
Description
Description
For use with vacuum pumps
Specifications
Specifications
Model M8C and M12C vacuum pump
Gasket set, exhaust valves, valve springs, lip seals, O-ring, motor coupling
Pump Repair Kit