missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Stainless Steel Shaft for Mixing Impellers

Stainless Steel Shaft for Mixing Impellers is composed of 316L stainless steel and passivated for maximum corrosion protection. Available in multiple sizes.

Supplier:  Fisherbrand™ A733FSR

 View more versions of this product

Catalog No. 14-500-368


Explore available promotions

Description

Description

  • Composed of stainless steel 316L
  • 9.5 mm (0.375 in.) shafts have a turned down 8 mm (0.3125 in.) end
  • Passivated for corrosion resistance
Specifications

Specifications

0.375 in.
Any mixer that accepts 9.5 mm shaft
508 mm
Stainless Steel (316L)
9.5 mm
20 in.
Impeller Stirrer Shaft
Videos
SDS
Documents

Documents

Accessories
Promotions

Promotions