Fisherbrand™ Stand and Clamp for Model 50 and 120 Sonic Dismembrator
Stand and Converter clamp accessory for Model 50/120 Sonic Dismembrator. This accessory will properly support the converter and probe assembly during sonication.
Supplier:  Fisherbrand™ FB460
Catalog No. FB460
Explore available promotions
Description
Description
This Fisher Scientific™ Stand is designed for use with Model 50 and Model 120 Sonic Dismembrators
  • Laboratory Support Stand with Converter clamp
  • For use with Model 50/120 Sonic Dismembrator
Specifications
Specifications
Stand
61 cm
0.5 in. dia. support rod and converter clamp
24 in.
Model 50 and 120 sonic dismembrator