Fisherbrand™ Tissue Path™ MACROSETTE™ Processing/Embedding Cassettes
Tissue Path™ MACROSETTE™ Processing/Embedding Cassettes for holding large tissue specimens during processing and embedding.
Catalog No. 15-182-706
Explore available promotions
Description
Description
  • Made from a special high-density acetal polymer
  • Efficient flow-through slots maximize fluid exchange and ensure proper drainage
  • One-piece design eliminates the need for separate lids
  • Cassette can be opened or closed as many times as needed
  • External L x W: 40.1 x 28.5 mm (1.58 x 1.1 in.)
  • For large specimens
Specifications
Specifications
White
13 mm
28.5 mm
Processing Cassette
0.5 in.
40.1 mm
Acetal