missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Traceable™ Clock/Thermometer/Humidity Meter

Battery-operated, wall-mounted unit has 9.75 in. dial

Catalog No. 06-664-10


Explore available promotions

Includes: AA battery and screw for wall mounting

Description

Description

Keep track of the current time, temperature and humidity all at once with the Fisher Scientific™ Traceable™ Clock/Thermometer/Humidity Meter. Battery-operated, wall-mounted unit has 9.75 in. dial.

  • Display of time to the second with 0.01% quartz crystal accuracy
  • Temperature: -20° to +55°C (-4° to +131°F)
  • Relative humidity: 0 to 100%

Certifications

An individually serial-numbered Traceable™ ISO 17025 certificate is provided to indicate instrument traceability (for time only) to standards provided by NIST.

Specifications

Specifications

Clock Thermometer/Humidity Thermometer
0.02
-20°C to +55°C
10°F to 130°F
1 lb.
Keep track of the current time, temperature and humidity all at once
ISO 17025, NIST
0 to 100%
±5%
±1.5°C
2°F
0.453 kg
AA battery and wall mount screw
Videos
SDS
Documents

Documents

Promotions

Promotions