Fisherbrand™ Traceable™ Digital Thermometer
The laboratory standard
Catalog No. 15-077-8
Explore available promotions
Includes: Stainless steel probe; 9V Alkaline battery; High-impact, chemical-resistant ABS plastic case
Description
Description
Provides routine measurements with semisolid, liquid, or air/gas. Fisherbrand™ Traceable™ Digital Thermometers can switch between Celsius and Fahrenheit temperatures. Easy-to-read LCD screen includes Min/Max memory. Switch-selectable temperature ranges: -50° to +150°C and -58° to +302°F.
  • Universal, stainless-steel 316 probe is triple purpose for liquid, air/gas, and semisolids.
  • Ideal for foods, pharmaceuticals, petroleum products, gas systems, soil, ovens, and more
  • Unit reads in °C and °F and includes a HOLD key for freezing data on the display for future readings
  • Instant HISTORY key recalls minimum and maximum readings over the past 24 hours
  • Unit conforms to ITS-90 International Temperature Standard
Order Info
Order additional probes separately; contact your Customer Service Representative. Accessory adapter also available.
Certifications
Individually serial-numbered Traceable Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™; indicates traceability to standards provided by NIST™.
Specifications
Specifications
ISO 17025, A2LA, NIST
32 mm
Measuring temperatures in cuvettes, gas systems, reagents, chemical solutions, soil, water baths, ovens, incubators, pharmaceuticals, petroleum products, foods and wastewater
140 mm
59 in.
150 cm
ABS Plastic
0.0001°C
226.8 g
9 in.
150°C
-40°C
-40°C to +150°C
Digital LCD
1.26 in.
0.5 in.H (13 mm)
5.5 in.
Stainless steel probe with USB connector; 9V Alkaline battery; High-impact, chemical-resistant ABS plastic case
6 in.
152.4 mm
Stainless Steel with USB Connector
8 oz.
3.5 in.
300°F
-40°F
-40°F to +300°F
±0.05°C between 0°C to 100°C
89 mm
Videos
SDS
Documents
Documents
Accessories