Fisherbrand™ Traceable™ Digital Thermometers with Short Sensors
Fisherbrand™ Traceable™ Digital Thermometers are ideal for monitoring liquids, air/gas or semisolids.
 View more versions of this product
Catalog No. 15-077-20
Explore available promotions
Description
Description
Choose from models with a variety of features including MAX/MIN function to record the maximum and minimum temperature in a certain time period; visual and audible alarms and multiple displays of recorded temperatures.
  • Select the correct temperature to use in water baths, refrigerators, incubators and tanks
Certifications
Individually serial-numbered Traceable Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™ indicates traceability to standards provided by NIST™.
Specifications
Specifications
ISO/IEC 17025:2005 (1750.01), A2LA
0.5 cm
0.75 in. (1.9 cm) H
0.5 in.
Solar panels, backup battery for dim areas, external sensor
102 mm
Updates every 10 sec.
2 oz.
1.75 in.
4 in.
70°C
-50°C
-50°C to +70°C
Digital LCD
0.19 in.
4 x 1.75 x 0.5 in. (10.2 x 4.4 x 1.3 cm)
Measuring temperatures of solutions in water baths, refrigerators, incubators and tanks
1.3 cm
10 ft.
304.8 cm
0.1°
56 g
Digital Thermometer: Dual Scale
158°F
-58°F
-58°F to +158°F
±1°C
4.4 cm
Videos
SDS
Documents
Documents
Accessories