Fisherbrand™ Traceable™ Dual-Range Light Meter
Measures both lux and foot-candles at a rapid sampling rate of 0.4 seconds. Display update rate is adjustable to 1 or 2 seconds.
Description
Measures both lux and foot-candles at a rapid sampling rate of 0.4 seconds. Display update rate is adjustable to 1 or 2 seconds.
- Lux measurements: three ranges from 0 to 1999, 2000 to 19,990 and 20,000 to 50,000 lux; resolution from 1 to 100 lux; accuracy of ±5% plus two digits
- Foot-candle measurements: three ranges from 0 to 199.9; 200 to 1999 and 2000 to 5000 ft-c.; resolution from 0.1 to 10 ft-c; accuracy of ±5% plus two digits
- Push-button access to highest, lowest and average of recorded readings
- DATA HOLD function “freezes” display while analog output allows readings to be captured by a recorder
- Photodiode and color-correction filter provide cosine- and color-corrected measurements
Order Info
Dimensions: 6.5L x 3W x 1.5 in. H (16.5 x 7.6 x 3.8cm). Net weight: 10 oz. (283g)
Certifications
Individually serial-numbered Traceable Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™. It indicates traceability to standards provided by NIST™.
Specifications
±5% plus two digits | |
Light Meter | |
3.8 cm | |
2 in. Diameter (5.1cm) photocell, 40 in. (1m) cable, 9V alkaline battery and recorder output | |
Dual Range | |
1 to 100 lux | |
Individually serial-numbered Traceable Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™. It indicates traceability to standards provided by NIST™. | |
0 to 199.9; 200 to 1999 and 2000 to 5000 ft-c |
6.5 in. | |
±5% plus two digits | |
0.1 to 10 ft-c | |
3 in. | |
7.62 cm | |
16.5 cm | |
0 to 1999, 2000 to 19,990 and 20,000 to 50,000 lux |