missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Traceable™ Excursion-Trac™ Datalogging Thermometer
Traceable™ Excursion-Trac™ Datalogging Thermometer with user-defined time intervals.
Description
Traceable™ Excursion-Trac™ Datalogging Thermometer with user-defined time intervals.
High-accuracy thermometer has a temperature range of -50 to 70°C and resolution of 0.01°C and maintains an accuracy of ±0.25°C across the entire temperature range.
- Stores up to ten unique alarm events
- Meets current CDC requirements for vaccine storage and monitoring
- Rolling memory structure, maintains most recent 525,600 temperature observations
- Memory can be cleared on unit after downloading data to USB stick
- Hassle-free retrieval of data; unit can remain in use while downloading and analyzing data
- Status indicators: Low battery, memory full, USB data transfer and active alarm state
- Traceable to NIST for accuracy
- High-impact, chemical-resistant ABS plastic case measures 7 x 10.8 x 1.9 cm (2.75 x 4.25 x 0.75 in.)
- Datalogging Capacity: 525,600 temperature observations
- Weight: 5 oz.
- Alarm tracking feature stores in memory each alarm event detailing when the alarm state occurred, as well as when the unit returned back to within range
- Alarm state indicators include visual LEDs, audio alerts, and flashing LCD segments
- Data for up to ten different alarm events can be retrieved with the most current event viewable on the display
- New feature allows recorded data (CSV file) to be transferred from thermometer to PC or Mac™ using a USB flash drive (not included)
- No software is needed for computer interface
- Each thermometer has a unique ID allowing multiple units to be used in the same location
- Monitor readings overnight, on weekends, or for any time period with rolling data log
- User-defined data-logging interval of one observation/minute to one observation/24 hours
- Memory capacity maintains a year of recorded data when using one-minute intervals and longer when using larger intervals
- Bottle probes are sealed in a miniature bottle (1 x 2.5 in.) filled with nontoxic glycol
- Solution is GRAS (generally recognized as safe) by the FDA (Food and Drug Administration)
- Eliminates concerns about incidental contact with food or drinking water
- Temperature-buffered bottle sensor eliminates rapid changes when refrigerator door is opened
- Ideal for use in monitoring storage conditions for vaccines or other important samples
Note: Units are designed to be used with either the provided AC adaptor or with batteries only. Batteries are designed to last for up to six months. If using battery-power only, the AC power adaptor must be plugged in to download any data.
Specifications
Datalogger | |
525,600 Readings | |
LED Indicator, Audible, Flashing LCD Segment | |
Stand, Velcro magnetic strips, wall mount, batteries | |
USB | |
±0.25°C | |
0.01° | |
-50.00°C to +70.00°C |
LCD | |
Calibrated | |
2 Probes (Bottle Probe and 5/16 in. dia. x 3/4 in. long) | |
ISO 17025, A2LA, NIST | |
Temperature-Buffered Bottle Sensor | |
0.01° | |
-58° to +158°F | |
10 ft. (Cable) |