missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Traceable™ Excursion-Trac™ Datalogging Thermometer

Traceable™ Excursion-Trac™ Datalogging Thermometer with user-defined time intervals.

 View more versions of this product

Catalog No. 15-081-128


Explore available promotions

Description

Description

Traceable™ Excursion-Trac™ Datalogging Thermometer with user-defined time intervals.

High-accuracy thermometer has a temperature range of -50 to 70°C and resolution of 0.01°C and maintains an accuracy of ±0.25°C across the entire temperature range.

  • Stores up to ten unique alarm events
  • Meets current CDC requirements for vaccine storage and monitoring
  • Rolling memory structure, maintains most recent 525,600 temperature observations
  • Memory can be cleared on unit after downloading data to USB stick
  • Hassle-free retrieval of data; unit can remain in use while downloading and analyzing data
  • Status indicators: Low battery, memory full, USB data transfer and active alarm state
  • Traceable to NIST for accuracy
  • High-impact, chemical-resistant ABS plastic case measures 7 x 10.8 x 1.9 cm (2.75 x 4.25 x 0.75 in.)
  • Datalogging Capacity: 525,600 temperature observations
  • Weight: 5 oz.
Hi/Lo Alarms and Time/Date Stamps
  • Alarm tracking feature stores in memory each alarm event detailing when the alarm state occurred, as well as when the unit returned back to within range
  • Alarm state indicators include visual LEDs, audio alerts, and flashing LCD segments
  • Data for up to ten different alarm events can be retrieved with the most current event viewable on the display
USB Data Transferring
  • New feature allows recorded data (CSV file) to be transferred from thermometer to PC or Mac™ using a USB flash drive (not included)
  • No software is needed for computer interface
  • Each thermometer has a unique ID allowing multiple units to be used in the same location
Data-Logging Features
  • Monitor readings overnight, on weekends, or for any time period with rolling data log
  • User-defined data-logging interval of one observation/minute to one observation/24 hours
  • Memory capacity maintains a year of recorded data when using one-minute intervals and longer when using larger intervals
Bottle Probe Features (Cat. Nos. 15-081-125, -126, -128, 129 do not come with bottle probes)
  • Bottle probes are sealed in a miniature bottle (1 x 2.5 in.) filled with nontoxic glycol
  • Solution is GRAS (generally recognized as safe) by the FDA (Food and Drug Administration)
  • Eliminates concerns about incidental contact with food or drinking water
  • Temperature-buffered bottle sensor eliminates rapid changes when refrigerator door is opened
  • Ideal for use in monitoring storage conditions for vaccines or other important samples

Note: Units are designed to be used with either the provided AC adaptor or with batteries only. Batteries are designed to last for up to six months. If using battery-power only, the AC power adaptor must be plugged in to download any data.

Specifications

Specifications

Datalogger
525,600 Readings
LED Indicator, Audible, Flashing LCD Segment
Stand, Velcro magnetic strips, wall mount, batteries
USB
±0.25°C
0.01°
-50.00°C to +70.00°C
LCD
Calibrated
1 Stainless-Steel Probe
ISO 17025, A2LA, NIST
Temperature-Buffered Bottle Sensor
0.01°
-58° to +158°F
10 ft. (Cable)
Videos
SDS
Documents

Documents

Promotions

Promotions