missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Traceable™ RTD Platinum Thermometer

High-precision, durable thermometer with platinum RTD probe, and linear fuzzy logic chip for wide temperature-range operation

Catalog No. 15-077-55


Explore available promotions

Includes: 9V battery and probe with 40 in. (102cm) cable

Description

Description

Platinum (100ohm) four-wire RTD probe can be used in liquids, air/gas, and semisolids.

  • Resolution: 0.01° below 200° and 0.1° above 200°
  • Accuracy: ±(0.1% + 0.2°C) below 200°, ±(0.15% + 0.5°C) above 200°
  • Probe range: -100° to +300°C
  • Features HOLD button to freeze display and capture reading
  • Fail-safe display indicates low-battery and probe condition
  • Easy-to-read, high-contrast LCD with 0.51 in.H (1.3cm) digits
  • Readings are updated 2.5 times every second
  • Ideal for experimental applications and in severe environments
  • Reproducible one-button operation minimizes operator error in quality control environments
  • Use to measure temperatures in soil, foods, ovens, cuvets, gas systems, reagents, water baths, wastewater, incubators, chemical solutions, petroleum products, and extreme cold
 

Order Info

Unit dimensions: 1.25L x 3W x 7 in.H (3.2 x 7.6 x 17.8cm); Weight: 9.5 oz. (269g); Probe measures 6 in.; Overall length: 12 in.

Certifications

Individually serial-numbered Traceable Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™; indicates traceability to standards provided by NIST™.

Specifications

Specifications

ISO 17025, A2LA, NIST
4 x 3 x 7 in. (10.1 x 7.6 x 17.8 cm)
7 in.
9 V Battery and probe with 40 in. (102cm) cable
101 mm
0.1°C
200 g
4 in.
300°C
-100°C
Points of calibration: 0, 50, and 100°C
Digital LCD
0.13 in.
Lab and plant use
17.8 cm
9 in.
22.8 cm
9.5 oz.
3 in.
572°F
-148°F
-100°C to +300°C
0.5°C
7.6 cm
Videos
SDS
Documents

Documents

Promotions

Promotions