Learn More
Fisherbrand™ Vortex Mixer Accessories: Foam Insert Kit
For use with Fisherbrand vortex mixers.
$80.79 - $128.55
Specifications
For Use With (Equipment) Vortex mixer
View More Specs
Products 2
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
02-215-396
View Documents
Fisherbrand™
80862991
Foam Insert Kit
Each for $128.55
Only null left
02-215-397
Accessories View Documents
Fisherbrand™
80863002
Blank Foam insert
Each for $80.79
Only null left
Description
Description
Also compatible with other manufacturers' models; contact your Fisher Scientific Customer Service Representative for details.
Foam Insert Variety Pack
  • Includes foam inserts for the following: 9 to 13mm tube holder, 14 to 19mm tube holder, 20 to 29mm
Specifications
Specifications
Vortex mixer
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'