Fisherbrand™ Vortex Mixer Accessories Kits
For use with Fisherbrand vortex mixers.
Supplier:  Fisherbrand™ 80863011
 View more versions of this product
Catalog No. 02-215-390
Explore available promotions
Description
Description
Also compatible with other manufacturers' models; contact your Fisher Scientific Customer Service Representative for details.
Foam Microtube and Microwell Plate Holder Kit with Retainer
  • Includes one microtube holder, one microwell plate holder, and one insert retainer (Cat. Nos. 02-215-386, -387, -391)
Foam Tube Holder Kit with Retainer
  • Iincludes 3 tube holders (for 9-13mm, 14-19mm, and 20-25mm tubes), one flat foam insert, two vessel harnesses and one insert retainer (Cat. Nos. 02-215-393, -394, -395, -397, -392, -391)
Microtube Kit:
  • Mixes (48) 0.25 to 2mL micro tubes; includes foam insert and insert retainer
Microwell Plate Kit:
  • For mixing 96-well plates or deep well blocks; includes foam insert and insert retainer
Specifications
Specifications
Microwell Plate Kit
Vortex mixer
For mixing 96-well plates/deep well blocks; includes foam insert and insert retainer