Fisherbrand™ 24-Hour Urine Container
Features easy-to-read graduations and a convenient carrying handle
Description
Features easy-to-read graduations and a convenient carrying handle
- Amber colored for light filtration
- Plastic cap has an extra-thick liner to prevent leakage
- Inert to normal preservatives
Specifications
24-Hour Urine Specimen Collection Container | |
Yes | |
Amber | |
40/Cs. |
Extra-thick Liner Cap | |
Polyethylene | |
Plastic cap with extra thick liner; Convenient carrying handle | |
3000 mL |