Fisherbrand™ 60mL DOA Temp-Strip Urine Specimen Containers
Ideal for collection of urine for drug screening
Supplier:  Fisherbrand™ 22610131
 View more versions of this product
Catalog No. 22-610-131
Explore available promotions
Description
Description
  • Sterile and nonsterile indicator tab helps assure product sterility and tamper evidence protection before use
  • Bio-Tite™ cap is dual-threaded for a leak-resistant seal
  • 100% Virgin medical-grade polypropylene for high clarity
  • Nonsterile label reads “Sealed for protection”; Sterile reads “Sterile, tighten the cap securely”
  • White cap
Compliance
Meet 95 kPA standards for transport
Specifications
Specifications
Urine Specimen Container
Sterile
White
Collection of Urine for drug screening
500/Cs.
Polypropylene
95 kPA
Dual-threaded Cap
ID label (reads sterile, tighten the cap securely) with liquid crystal thermometer strip (90 to 100°F), indicator tab
60 mL