missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Absorption Macro Rectangular Cells with PTFE Lid

Fit standard spectrophotometer cell holders

Supplier:  Fisherbrand™ 14958115

 View more versions of this product

Catalog No. 14-958-115



Explore available promotions

Description

Description

Fit standard spectrophotometer cell holders

  • All cells are 12.5mm wide
  • 1, 5 or 10mm path length rectangular cells
  • Available with a variety of window materials for wide range of assays
  • Available in four materials e.g. Quartz, Optical Quartz, Special Optical Glass and Optical Glass
  • Quartz windows for transmission from far ultraviolet through the infrared
  • Glass windows for routine work in ultraviolet region and visible region

Specifications

Specifications

Macro Absorption Cell
Quartz
10mm
1 mm
52 mm
12.5 mm
Rectangle
0.35 mL
PTFE
2/Pk., 4 Pk./Cs.
200 to 3500 nm
Standard Spectrophotometer Cell Holder
Transmission from far ultraviolet through the infrared
Videos
SDS
Documents

Documents

Promotions

Promotions