Fisherbrand™ Absorption Quartz Micro Rectangular Cells with PTFE Lid
Fit standard spectrophotometer cell holders
Supplier:  Fisherbrand™ 14958134
 View more versions of this product
Catalog No. 14-958-134
Explore available promotions
Description
Description
Fit standard spectrophotometer cell holders
  • All cells are 12.5mm wide
  • 10mm path length rectangular cells
  • Reduce reagent use with Micro cells
  • Quartz windows for transmission from far ultraviolet through the infrared
Specifications
Specifications
Micro Absorption Cell
Quartz
2mm
2/Pk., 8 Pk./Cs.
200 to 2500 nm
Standard Spectrophotometer Cell Holder
Transmission from far ultraviolet through the infrared
500 μL
PTFE
2 clear windows
10 mm
45 mm
12.5 mm
Rectangle