missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Absorption Quartz Semi Micro Rectangular Cells with PTFE Lid, w/PTFE lid

Fit standard spectrophotometer cell holders

Supplier:  Fisherbrand™ 14958125

 View more versions of this product

Catalog No. 14-958-125



Explore available promotions

Description

Description

Fit standard spectrophotometer cell holders

  • All cells are 12.5mm wide
  • 10mm path length rectangular cells
  • Reduce reagent use with Semimicro cells
  • Quartz windows for transmission from far ultraviolet through the infrared

Specifications

Specifications

Semi-micro Absorption Cell
Quartz
4mm
10 mm
45 mm
12.5 mm
Rectangle
1000 μL
PTFE
2/Pk., 8 Pk./Cs.
200 to 2500 nm
Standard Spectrophotometer Cell Holder
Transmission from far ultraviolet through the infrared
Videos
SDS
Documents

Documents

Promotions

Promotions